missing translation for 'onlineSavingsMsg'
Learn More

21-Hydroxypregna-1,4,16-triene-3,11,20-trione (>85%), TRC

Product Code. 30407326
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407326 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407326 Supplier Toronto Research Chemicals Supplier No. TRCH95221025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 21-Hydroxypregna-1,4,16-triene-3,11,20-trione
CAS 102447-86-9
Molecular Formula C21H24O4
Quantity 25 mg
Synonym 16-Dehydro 17-Dehydroxy Prednisone
InChI Formula InChI=1S/C21H24O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h6-9,14-15,19,22H,3-5,10-11H2,1-2H3/t14-,15-,19+,20-,21-/m0/s1
SMILES C[C@]12CC(=O)[C@H]3[C@H]([C@@H]1CC=C2C(=O)CO)CCC4=CC(=O)C=C[C@]34C
IUPAC Name (8S,9S,10R,13S,14S)-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,12,14,15-hexahydro-6H-cyclopenta[a]phenanthrene-3,11-dione
Molecular Weight (g/mol) 340.41
Formula Weight 340.1675
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.