Learn More
Hamilton™ 21 gauge, Kel-F Hub Needle, custom length (0.375 to 12 in), point style 5 or AS, 6/PK
Product Code. 11533284
Change view
Click to view available options
Needle Gauge:
21 G
Unit Size:
Pack of 6
Product Code. Needle Gauge unitSize
11533284 21 G Pack of 6
1 options
Product Code. 11533284 Supplier Hamilton™ Supplier No. 775212
Only null left
Add to basket
Add to basket
Kel-F hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2 in. / 51 mm. Kel-F hub (KF) needles are available in convenient six packs. Custom lengths and point styles are available. These needles are available from 10 - 31 gauge.
Specification Sheet
  • Gauge – 21 gauge
  • Point Style – 5 or AS
  • Needle Length – 0.75 inch (19.1 mm)
  • Autoclavable – Yes
  • Gas Sterilizable – Yes
  • Sales Quantity – 6 needles per pack
  • Hub – Kel-F
Specifications
Needle Gauge 21 G
Product Type Kel-F Hub Needle