missing translation for 'onlineSavingsMsg'
Learn More

21-Desmethyl (17Beta)-21-Acetyloxy-4-chloro-17-methyl-18-norpregna-4,13-dien-3-one, TRC

Product Code. 30354171
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354171 25 mg 25mg
30355019 5 mg 5mg
30357779 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354171 Supplier Toronto Research Chemicals Supplier No. TRCD29355025MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 21-Desmethyl (17beta)-21-Acetyloxy-4-chloro-17-methyl-18-norpregna-4,13-dien-3-one
Analyte or Component Names 21-Desmethyl (17beta)-21-Acetyloxy-4-chloro-17-methyl-18-norpregna-4,13-dien-3-one
Shipping Condition Room Temperature
Molecular Formula C22H29ClO3
InChI Formula InChI=1S/C22H29ClO3/c1-13(24)26-12-21(2)10-8-15-14-4-5-18-20(23)19(25)9-11-22(18,3)17(14)7-6-16(15)21/h14,17H,4-12H2,1-3H3/t14-,17-,21?,22?/m0/s1
Solution Type Neat
SMILES [H][C@@]12CCC3=C(Cl)C(CC[C@]3(C)[C@@]1([H])CCC4=C2CC[C@]4(C)COC(C)=O)=O
IUPAC Name ((8R,9S,10R,17S)-4-chloro-10,17-dimethyl-3-oxo-2,3,6,7,8,9,10,11,12,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl)methyl acetate
Industry Type Forensics
Molecular Weight (g/mol) 376.92
Quantity 25 mg
Formula Weight 376.17
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.