Learn More
VELP Scientifica™ 21 CFR Part 11 Package for UDK 169/159 Distillation Units
Software upgrade for UDK 159 or 169 Distillation Units for compliant nitrogen determination according to FDA regulations.
Brand: VELP Scientifica™ A00000429
Additional Details : Weight : 0.01000kg
Description
This package helps achieve full compliance with Subpart B - Electronic Records and Subpart C - Electronic Signatures of the regulation:
- Track and record settings and any changes to settings with the system log function running permanently, serving as an audit trail
- Track who performs any operation as analysis results are automatically signed with the user information (each user has a unique identification and electronic signature)
- Ensure the quality and incorruptibility of recorded data with backup procedures (exported files are protected from unwanted or improper alteration)
- Ensure the right delegation of responsibilities at the right level with the user management system with three access levels
Specifications
Software Package | |
UDK 169 or 159 Distillation Units |