missing translation for 'onlineSavingsMsg'
Learn More

21-Carboxylic Acid Triamcinolone Acetonide, TRC

Product Code. 30008011
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008011 10 mg 10mg
30004853 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008011 Supplier Toronto Research Chemicals Supplier No. TRCC17855010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53962-41-7
Molecular Formula C24 H29 F O7
Molecular Weight (g/mol) 448.48
Synonym Pregna-1,4-dien-21-oic acid, 9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-3,20-dioxo-, (11β,16α)-, (11β,16α)-9-Fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-3,20-dioxopregna-1,4-dien-21-oic acid
IUPAC Name 2-[(1S,2S,4R,8S,9S,11S,12R,13S)-12-fluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-8-yl]-2-oxoacetic acid
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H29FO7/c1-20(2)31-17-10-15-14-6-5-12-9-13(26)7-8-21(12,3)23(14,25)16(27)11-22(15,4)24(17,32-20)18(28)19(29)30/h7-9,14-17,27H,5-6,10-11H2,1-4H3,(H,29,30)/t14-,15-,16-,17+,21-,22-,23-,24-/m0/s1
Formula Weight 448.1897
Chemical Name or Material 21-Carboxylic Acid Triamcinolone Acetonide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.