missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TUB Recombinant Rabbit Monoclonal Antibody (24GB1795)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557298
This item is not returnable.
View return policy
Description
TUB Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a member of the Tubby family of bipartite transcription factors. The encoded protein may play a role in obesity and sensorineural degradation. The crystal structure has been determined for a similar protein in mouse, and it functions as a membrane-bound transcription regulator that translocates to the nucleus in response to phosphoinositide hydrolysis. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| TUB | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TUB | |
| CELE_F10B5.4; F10B5.4; Rd5; RDOB; retinal degeneration 5; TUB; TUB bipartite transcription factor; TUB-1; tubby bipartite transcription factor; tubby candidate gene; tubby homolog; tubby homologue; tubby protein; Tubby protein homolog; Tubby protein homolog 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22141, 25609, 7275 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1795 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88808, P50586, P50607 | |
| TUB | |
| A synthesized peptide derived from human TUB1 (aa 1-50). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction