missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NPY5R Recombinant Rabbit Monoclonal Antibody (24GB60)

Product Code. 30276161
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30276161 20 μL 20µL
30275153 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30276161 Supplier Invitrogen™ Supplier No. MA557114

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NPY5R Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Receptor for neuropeptide Y and peptide YY. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity. Seems to be associated with food intake. Could be involved in feeding disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPY5R
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 24GB60
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Npy5r
Gene Accession No. O70342, Q15761, Q63634
Gene Alias HGNC:7958; Neuropeptide Y receptor type 5; neuropeptide Y receptor type 5-like protein; neuropeptide Y receptor Y5; Neuropeptide Y5 receptor; Npy5; NPY5 receptor; Npy5r; NPY5-R; NPYR5; NPYY5; NPY-Y5 receptor; NPYY5-R; Y5 receptor; Y5R
Gene Symbols Npy5r
Host Species Rabbit
Immunogen A synthesized peptide derived from human NPY5R (aa 1-28).
Purification Method Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18168, 25340, 4889
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.