missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCTD9 Recombinant Rabbit Monoclonal Antibody (23GB6305), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553017
This item is not returnable.
View return policy
Description
Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex, which mediates the ubiquitination of target proteins, leading to their degradation by the proteasome.
Specifications
| KCTD9 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Kctd9 | |
| BTB/POZ domain-containing protein KCTD9; BTBD27; KCTD9; potassium channel tetramerisation domain containing 9; potassium channel tetramerization domain containing 9 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54793 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6305 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7L273 | |
| Kctd9 | |
| A synthesized peptide derived from human KCTD9 (61-85AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction