missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMAL1 Recombinant Rabbit Monoclonal Antibody (23GB4875), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552769
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| BMAL1 | |
| Recombinant Monoclonal | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00327, Q9EPW1, Q9WTL8 | |
| Arntl | |
| A synthesized peptide derived from human BMAL1 (555-600AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4875 | |
| Unconjugated | |
| Arntl | |
| Arnt3; ARNTL; ARNT-like protein 1, brain and muscle; aryl hydrocarbon receptor nuclear translocator like; aryl hydrocarbon receptor nuclear translocator-like; aryl hydrocarbon receptor nuclear translocator-like protein 1; basic-helix-loop-helix-PAS orphan MOP3; basic-helix-loop-helix-PAS protein MOP3; bHLHe5; bHLH-PAS protein JAP3; BMAL1; Bmal1 protein; BMAL1b; bmal1b'; BMAL1c; Brain and muscle ARNT-like 1; brain and muscle Arnt-like protein 1; CG8727 PA; class E basic helix-loop-helix protein 5; cycle; JAP3; LOW QUALITY PROTEIN: aryl hydrocarbon receptor nuclear translocator-like protein 1; member of PAS protein 3; member of PAS superfamily 3; MGC47515; MOP3; PAS domain-containing protein 3; PASD3; testis tissue sperm-binding protein Li 50e; tic | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11865, 29657, 406 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction