missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TMPRSS2 His-tag Protein, CF
Description
Measured by its ability to cleave the fluorogenic peptide substrate Boc-QAR-AMC (Catalog # ES014 ). The specific activity is >7000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | NP_005647.3 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in MES and NaCl. |
| Gene ID (Entrez) | 7113 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 19-25 kDa (non-catalytic) & 25-33 kDa (catalytic) under reducing conditions, MolecularWeight-theroretical: 17 kDa (non-catalytic), 29 kDa (catalytic) |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human TMPRSS2 protein Trp106-Gly492, with modifications in the non-catalytic chain and a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70°C as supplied. 3 months, -20 to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | EC 3.4.21, Epitheliasin, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?