missing translation for 'onlineSavingsMsg'
Learn More

(20R)-Protopanaxadiol, MedChemExpress

Product Code. 16419258
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16419258

Brand: Medchem Express HYN2040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (20R)-Protopanaxadiol is a triterpenoid saponin metabolite of 20(R)-ginsenoside Rg3 in black ginseng. (20R)-Protopanaxadiol exhibits anti-tumor activity and cytotoxicity, and potently inhibits the growth of Helicobacter pylori.

(20R)-Protopanaxadiol is a triterpenoid saponin metabolite of 20(R)-ginsenoside Rg3 in black ginseng. (20R)-Protopanaxadiol exhibits anti-tumor activity and cytotoxicity, and potently inhibits the growth of Helicobacter pylori.

Specifications

Chemical Name or Material (20R)-Protopanaxadiol
CAS 7755-01-3
Color White
Purity Grade Notes Research
Molecular Formula C30H52O3
Quantity 10 mg
Solubility Information DMSO : 20 mg/mL (43.41 mM; Need ultrasonic)
SMILES C[C@]([C@@](C[C@H]1O)([H])[C@]2(CC[C@@H]3O)C)(CC[C@@]2([H])C3(C)C)[C@]4([C@@]1([H])[C@]([C@](C)(O)CC/C=C(C)/C)([H])CC4)C
Molecular Weight (g/mol) 460.73
Formula Weight 460.73
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.