missing translation for 'onlineSavingsMsg'
Learn More

20Alpha-Hydroxy-5Alpha-pregnan-3-one Dimethyl Acetal, TRC

Product Code. 30395360
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395360 25 mg 25mg
30392759 100 mg 100mg
30391207 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395360 Supplier Toronto Research Chemicals Supplier No. TRCH94737025MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 20alpha-Hydroxy-5alpha-pregnan-3-one Dimethyl Acetal
Analyte or Component Names 20alpha-Hydroxy-5alpha-pregnan-3-one Dimethyl Acetal
Shipping Condition Room Temperature
Molecular Formula C23H40O3
InChI Formula InChI=1S/C23H40O3/c1-15(24)18-8-9-19-17-7-6-16-14-23(25-4,26-5)13-12-21(16,2)20(17)10-11-22(18,19)3/h15-20,24H,6-14H2,1-5H3/t15-,16-,17-,18+,19-,20-,21-,22+/m0/s1
Solution Type Neat
SMILES C[C@@]12[C@](CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@@H](O)C)([H])CC(OC)(OC)CC1
IUPAC Name (S)-1-((5S,8R,9S,10S,13S,14S,17S)-3,3-dimethoxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)ethan-1-ol
Industry Type Forensics
Molecular Weight (g/mol) 364.56
Quantity 25 mg
Formula Weight 364.3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.