missing translation for 'onlineSavingsMsg'
Learn More

20Alpha-Dihydro Pregnenolone-d5, TRC

Product Code. 30387780
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387780 10 mg
30374735 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30387780 Supplier Toronto Research Chemicals Supplier No. TRCD44640210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C21H29D5O2
Molecular Weight (g/mol) 323.52
Synonym (3β,20S)-Pregn-5-ene-3,20-diol-d5, Pregn-5-ene-3β,20α-diol-d5, Pregnenediol-d5, (3β,20S)-Pregn-5-ene-3,20-diol-d5, 20α-Hydroxypregnenolone-d5, 3β,20α-Dihydroxypregn-5-ene-d5, Δ5-Pregnene-3β,20α-diol-d5, Δ5-Pregnenediol-d5
IUPAC Name (3S,8S,9S,10R,13S,14S,17S)-17-((S)-1-hydroxyethyl-1,2,2,2-d4)-10,13-dimethyl-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-d-3-ol
SMILES [H][C@@]12CC=C3C[C@H](CC[C@@]3([C@]1(CC[C@]4([C@]2(CC[C@@]4([C@](C([2H])([2H])[2H])(O)[2H])[2H])[H])C)[H])C)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H34O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4,13,15-19,22-23H,5-12H2,1-3H3/t13-,15-,16-,17+,18-,19-,20-,21+/m0/s1/i1D3,13D,17D
Formula Weight 323.29
Chemical Name or Material 20alpha-Dihydro Pregnenolone-d5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.