missing translation for 'onlineSavingsMsg'
Learn More

20alpha-Dihydro Pregnenolone 3-Sulfate Sodium Salt, TRC

Product Code. 30380436
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380436 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380436 Supplier Toronto Research Chemicals Supplier No. TRCD44640525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 20alpha-Dihydro Pregnenolone 3-Sulfate Sodium Salt
CAS 131320-06-4
Molecular Formula C21 H33 O5 S . Na
Quantity 25 mg
Synonym (3β,20S)-Pregn-5-ene-3,20-diol 3-(Hydrogen Sulfate) Sodium, Pregn-5-ene-3β,20α-diol 3-(Hydrogen Sulfate) Sodium, 5-Pregnene-20α-ol 3β-Sulfate Sodium, Dihydropregnenolone 3-Sulfate Sodium, Pregnenediol Sulfate Sodium
InChI Formula InChI=1S/C21H34O5S.Na/c1-13(22)17-6-7-18-16-5-4-14-12-15(26-27(23,24)25)8-10-20(14,2)19(16)9-11-21(17,18)3;/h4,13,15-19,22H,5-12H2,1-3H3,(H,23,24,25);/q;+1/p-1/t13-,15+,16-,17?,18-,19-,20-,21+;/m0./s1
SMILES [Na+].C[C@H](O)C1CC[C@H]2[C@@H]3CC=C4C[C@@H](CC[C@]4(C)[C@H]3CC[C@]12C)OS(=O)(=O)[O-]
IUPAC Name sodium;[(3R,8S,9S,10R,13S,14S)-17-[(1S)-1-hydroxyethyl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] sulfate
Molecular Weight (g/mol) 420.538
Formula Weight 420.195
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.