missing translation for 'onlineSavingsMsg'
Få mere at vide

LMS™ Series 3 Model 200WNP Cooled Incubator, 227 L, Stainless Steel

Produktkode 11837592
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
227 L
Förpackningsstorlek
Each
Denne vare kan ikke returneres. Se returpolitik

Produktkode 11837592

missing translation for 'mfr': LMS™ 200NP

Kun null tilbage
Tilføj til indkøbskurv
Tilføj til indkøbskurv
Denne vare kan ikke returneres. Se returpolitik

Temperature Range: -10°C to +50°C

  • Range of mid-sized benchtop and freestanding cooled incubators with a white stove enamel mild steel exterior, foam insulation and stainless steel interiors
  • Reliable and accurate temperature control
  • Standard single and dual temperature, plus programmable models available
  • Temperature range -10.0°C to +50.0°C, temperature variation with average workload±0.5°C
  • Self-evaporating condensate drip tray
  • White plastic coated wire shelves
  • Energy efficient control system and eco-friendly
  • P.I.D. heating and cooling to ensure stable internal temperature
  • Pre-set safety cutout
  • Hermetically sealed refrigeration system
  • Door lock
  • Digital controls are easy to read and simple to use
  • Full range offactory-fitted optional extras available, including LED lighting, access porthole, alarm system, inner perspex door, internal socket and chart recorder
  • Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests

Specifications

Certifications/Compliance ISO 9001:2008
Controller Type Programmable
Description Freestanding incubator
Capacity (Metric) 227 L
Chamber Material Stainless Steel
Convection Type Fan-assisted Circulation
Dimensions (D x W) Shelf 510 mm x 390 mm
Lighting Optional Fluorescent/UV
Material Stove Enameled Mild Steel
No. of Shelves 3
Temperature Range (Metric) -10°C to +50°C
Type Refrigerated Incubator
Display Digital
Insulation Foamed polyurethane
Model 200WNP
No. of Doors 1
Refrigerant CFC-free
Shelf Area (Metric) 510 x 390 mm
Temperature Stability ±0.1°C
Temperature Uniformity ±0.5°C
Warranty 2 Years
Width (English) 26.8 in.
Width (English) Interior 21.6 in.
Width (Metric) 680 mm
Width (Metric) Interior 550 mm
Height (English) 61.0 in.
Height (English) Interior 36.2 in.
Height (Metric) 1550 mm
Height (Metric) Interior 920 mm
Depth (English) Interior 17.7 in.
Depth (Metric) Interior 450 mm
Weight (Metric) 125 kg
Electrical Requirements 240 V, 50 Hz
Voltage 230 V
Frequency 50 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.