missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ganglioside GM2 Chimeric Recombinant Rabbit Monoclonal Antibody (KM966)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551582
This item is not returnable.
View return policy
Description
Specificity: This antibody recognizes and binds ganglioside GM2.
Ganglioside GM2 is a glycosphingolipid found in the plasma membrane of cells, particularly in the nervous system. It is composed of a ceramide lipid tail and a complex carbohydrate head group that contains sialic acid residues. The function of Ganglioside GM2 is not fully understood, but it is believed to play a role in cell signaling and communication, as well as in the formation and maintenance of the myelin sheath that surrounds nerve cells. Mutations in the genes responsible for the synthesis or degradation of Ganglioside GM2 can result in various neurological disorders, including Tay-Sachs disease and Sandhoff disease.
Specifications
| Ganglioside GM2 Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| This chimeric antibody was generated by cloning the variable heavy and light chain sequences of murine parent antibody KM696 (IgM) and expressing it as a mouse/human IgG1. The parent antibody was raised against human GM2. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry | |
| KM966 | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction