missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta Galactosidase Chimeric Recombinant Mouse Monoclonal Antibody (13R4)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551772
This item is not returnable.
View return policy
Description
13R4 recognizes wild-type E. coli beta-galactosidase.
Beta Galactosidase encodes a member of the glycosyl hydrolase 35 family of proteins. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed to generate the mature lysosomal enzyme. This enzyme catalyzes the hydrolysis of a terminal beta-linked galactose residue from ganglioside substrates and other glycoconjugates. Mutations in this gene have been associated with GM1-gangliosidosis and Morquio B syndrome. Diseases associated with GLB1 include Gm1-Gangliosidosis, Type I and Gm1-Gangliosidosis, Type Ii.
Specifications
| beta Galactosidase Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Mouse | |
| Metal-chelate Chromatography | |
| RUO | |
| Bacteria | |
| Antibody | |
| F(ab) λ |
| Flow Cytometry, Functional Assay, Western Blot | |
| 13R4 | |
| Unconjugated | |
| acid beta-galactosidase; AW125515; betagal; Beta-Gal; Beta-galactosidase; bgal; Bge; Bgl; Bgl-e; Bgl-s; Bgl-t; Bgs; Bgt; C130097A14Rik; EBP; Elastin receptor 1; elastin receptor 1, 67kDa; ELNR1; galactosidase beta 1; galactosidase, beta 1; GLB1; Glb-1; lactase; lysosmal hydrolase; lysosomal beta-galactosidase; lysosomal hydrolase; MPS4B; βgal; β-gal; β-galactosidase | |
| Beta-galactosidase. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction