missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IgG1, Alexa Fluor 405, Isotype Control, Clone: 43414, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems IC005V
This item is not returnable.
View return policy
Description
IgG1 Monoclonal specifically detects IgG1 in Rat samples. It is validated for Control.
Specifications
| IgG1 | |
| 43414 | |
| Control 5 uL/10^6 cells | |
| IgG1, Igh-4, RGD1359539, VH7183 | |
| KLH | |
| 200 Tests | |
| Primary | |
| Rat | |
| Purified |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Selected for use as rat IgG 1 isotype control for flow cytometry applications. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction