missing translation for 'onlineSavingsMsg'
Learn More

Rapamycin, MedChemExpress

Product Code. 16440956
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
Unit Size:
100mg
1g
200mg
2g
500mg
50mg
5g
This item is not returnable. View return policy

Product Code. 16440956

Brand: Medchem Express HY10219

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rapamycin (Sirolimus; AY 22989) is a potent and specific mTOR inhibitor with an IC50 of 0.1 nM in HEK293 cells. Rapamycin binds to FKBP12 and specifically acts as an allosteric inhibitor of mTORC1. Rapamycin is an autophagy activator, an immunosuppressant.

Rapamycin (Sirolimus

Specifications

Chemical Name or Material Rapamycin
CAS 53123-88-9
Color White
Purity Grade Notes Research
Molecular Formula C51H79NO13
Quantity 200 mg
Synonym Sirolimus AY-22989
Solubility Information DMSO : 125 mg/mL (136.74 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (54.69 mM; Need ultrasonic)
SMILES O=C([C@@]1(O)[C@@H](CC[C@@H](C[C@@H](/C(C)=C/C=C/C=C/[C@H](C[C@@H](C)C([C@@H]([C@@H](/C(C)=C/[C@H]2C)O)OC)=O)C)OC)O1)C)C(N3CCCC[C@H]3C(O[C@@H](CC2=O)[C@@H](C[C@H]4C[C@H]([C@H](O)CC4)OC)C)=O)=O
Molecular Weight (g/mol) 914.17
Formula Weight 914.17
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.