missing translation for 'onlineSavingsMsg'
Learn More

Polyoxyethylene stearate, MedChemExpress

Product Code. 16400936
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
Unit Size:
1g
200mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16400936 200 mg 200mg
16480769 1 g 1g
16406137 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16400936 Supplier Medchem Express Supplier No. HY101530

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

MedChemExpress Polyoxyethylene stearate (POES) is a non-ionic emulsifying agent.

Polyoxyethylene stearate (POES) is a non-ionic emulsifying agent.

Specifications

Chemical Name or Material Polyoxyethylene stearate
Physical Form Solid
CAS 9004-99-3
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C20H40O3 (monomer)
For Use With (Application) Neuroscience-Neuromodulation
Synonym POES
Solubility Information DMSO : 100 mg/mL (Need ultrasonic)
SMILES O=C(OCCO)CCCCCCCCCCCCCCCCC.[n=10].[n]
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.