Learn More
Fisher Chemical 200LT N-HEXANE - CERTIFIED REAGENTBE USED ASA MOTOR FUEL,HEATING FUEL ADDITIVE OR
Brand:  Fisher Chemical H/0421/27
200LT N-HEXANE - CERTIFIED REAGENTBE USED ASA MOTOR FUEL,HEATING FUEL ADDITIVE OR
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 12025470
  • ONLINE EXCLUSIVE PRICE
    £15055.00
    £14426.00 / 200 L
    Save £629.00
    4% Off
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361f-Suspected of damaging fertility.
  • H336-May cause drowsiness or dizziness.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P273-Avoid release to the environment.