Learn More
Fisher Chemical 200LT Heptane, extra pure, fraction, SLR
Brand:  Fisher Chemical H/0100/27
200LT Heptane, extra pure, fraction, SLR
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11498323
  • ONLINE EXCLUSIVE PRICE
    £3090.00
    £2943.00 / 200 L
    Save £147.00
    4% Off
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Heptane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P331-Do NOT induce vomiting.
  • P273-Avoid release to the environment.