Learn More
200LT CYCLOHEXANE - CERTIFIED REAGENTOTHER THAN AS HEATING FUEL,MOTOR FUEL,OR ADDITIVE
Brand: Fisher Chemical C/8921/27
200LT CYCLOHEXANE - CERTIFIED REAGENTOTHER THAN AS HEATING FUEL,MOTOR FUEL,OR ADDITIVE
Code : 27
Additional Details : CAS Number : 110-82-7 Weight : 154.00000kg Transport : UN number : 1145 Chem class : 3 Pack group : II
Safety and Handling
- Cyclohexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.