missing translation for 'onlineSavingsMsg'
Learn More

Fisher Chemical 200LT ACETONITRILE - SPECIFIED REAGENT

Brand:  Fisher Chemical A/0620/27


200LT ACETONITRILE - SPECIFIED REAGENT

Product Code. 10326182

  • ONLINE EXCLUSIVE PRICE
    £3595.00
    £3307.00 / 200 L
    Save £288.00
    8% Off

Please to purchase this item. Need a web account? Register with us today!


For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Acetonitrile

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.