missing translation for 'onlineSavingsMsg'
Learn More

Zirconium Standard: Zr @ 1000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 17964376
Click to view available options
Quantity:
50 g
200 g
Unit Size:
200g
50g
This item is not returnable. View return policy

Product Code. 17964376

Brand: VHG Labs VHGOZR1000200G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Zirconium Standard: Zr @ 1000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zirconium
CAS 7440-67-7
Methods ICP/XRF/RDE
Analyte or Component Names Zirconium
Shipping Condition Room Temperature
Molecular Formula Zr
Synonym Zr
InChI Formula InChI=1S/Zr
SMILES [Zr]
IUPAC Name zirconium
Solvent or Matrix 75 cSt Mineral Oil
Molecular Weight (g/mol) 91.224
Quantity 200 g
Formula Weight 89.905 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.