missing translation for 'onlineSavingsMsg'
Learn More

Human IFN-beta Antibody, R&D Systems™

Product Code. 18706187
Change view
Click to view available options
Quantity:
20,000 U
Unit Size:
20000 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18706187 20,000 U 20000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18706187 Supplier R&D Systems Supplier No. 314011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IFN-beta Polyclonal specifically detects IFN-beta in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-beta
Applications Neutralization
Classification Polyclonal
Conjugate Unconjugated
Dilution Neutralization
Formulation Neat serum. with No Preservative
Gene Alias Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Gene Symbols IFNB1
Host Species Sheep
Immunogen Recombinant human IFN-beta 1a (expressed in mammalian cells)
Purification Method N/A
Quantity 20,000 U
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3456
Test Specificity Detects human IFN-beta .
Target Species Human
Content And Storage After receipt, the product may be stored at -20degreesC for short-term use (< 6 months). For long-term storage, we recommend storing the product at -70degreesC or below for retention of full activity. When thawing, the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of the product should be in buffers containing protein such as 0.1% bovine serum albumin (BSA).
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.