missing translation for 'onlineSavingsMsg'
Learn More

2000 μg/g Ethylene Glycol Standard in Multi-grade Diesel Engine Oil, VHG™

Product Code. 17957984
Click to view available options
Quantity:
80 mL
Unit Size:
80mL
This item is not returnable. View return policy

Product Code. 17957984

Brand: VHG Labs VHGEGOIL200080

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

2000 μg/g Ethylene Glycol Standard in Multi-grade Diesel Engine Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylene glycol
CAS 107-21-1
Analyte or Component Names Ethylene glycol
Shipping Condition Room Temperature
Molecular Formula C2 H6 O2
Concentration 2000 μg/g
Synonym Hydroxyethyl Salicylate Imp. B (EP), Ethylene Glycol, Propan-1,2-diol, Glycerol Impurity B, Hydroxyethyl Salicylate Impurity B
InChI Formula InChI=1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
SMILES OCCO
IUPAC Name ethane-1,2-diol
Solvent or Matrix Multi-grade Diesel Engine Oil
Molecular Weight (g/mol) 62.07
Quantity 80 mL
Formula Weight 62.0368 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.