missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ 200 mM 3-Aminobenzamide
Click to view available options
Quantity:
60 μL
Unit Size:
60µL
Description
Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Specifications
Specifications
| Content And Storage | Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 60 μL |
| Target | PARP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction