Learn More
Fisher Chemical 200 LT DICHLOROMETHANE HPLC IN STAINLESS STEELDRUM Please note a drum deposit of £285.00 will
Brand:  Fisher Chemical D/1856/27SS
200 LT DICHLOROMETHANE HPLC IN STAINLESS STEELDRUM Please note a drum deposit of £285.00 will
Product Code. 11493633
Request Bulk or Custom Formats
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.