missing translation for 'onlineSavingsMsg'
Learn More

200.8 Calibration Standard B: Ag, Ba @ 10 μg/mL in 1% HNO3, VHG™

Product Code. 17911696
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911696 500 mL 500mL
17922126 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17911696 Supplier VHG Labs Supplier No. VHGLSSB500
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

200.8 Calibration Standard B: Ag, Ba @ 10 μg/mL in 1% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium/Silver
CAS 7440-39-3, 7440-22-4
Methods ICP/ICP-MS
Analyte or Component Names Barium/Silver
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.