missing translation for 'onlineSavingsMsg'
Learn More

200.7, 6010 - Interference Check Solution 1: Hg @ 50; Be @ 100; Mn @ 200; Ag, Ba, Cd, Co, Cr, Cu, Ni, V, Zn @ 300; Se @ 500; As, Pb, Tl @ 1000; K @ 20000 μg/mL in 5% HNO3, VHG™

Product Code. 30003081
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003081 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003081 Supplier VHG Labs Supplier No. VHGINT1100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

200.7, 6010 - Interference Check Solution 1: Hg @ 50; Be @ 100; Mn @ 200; Ag, Ba, Cd, Co, Cr, Cu, Ni, V, Zn @ 300; Se @ 500; As, Pb, Tl @ 1000; K @ 20000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mercury/Beryllium/Manganese/Barium/Cadmium/Chromium/Cobalt/Copper/Nickel/Silver/Vanadium/Zinc/Selenium/Arsenic/Lead/Thallium/Potassium
CAS 7439-97-6, 7440-41-7, 7439-96-5, 7440-39-3, 7440-43-9, 7440-47-3
Methods ICP
Analyte or Component Names Mercury/Beryllium/Manganese/Barium/Cadmium/Chromium/Cobalt/Copper/Nickel/Silver/Vanadium/Zinc/Selenium/Arsenic/Lead/Thallium/Potassium
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Mercury 50 μg/mL/Beryllium 100 μg/mL/Manganese 200 μg/mL/Barium 300 μg/mL/Cadmium 300 μg/mL/Chromium 300 μg/mL/Cobalt 300 μg/mL/Copper 300 μg/mL/Nickel 300 μg/mL/Silver 300 μg/mL/Vanadium 300 μg/mL/Zinc 300 μg/mL/Selenium 500 μg/mL/Arsenic 1000 μg/mL/Lead 1000 μg/mL/Thallium 1000 μg/mL/Potassium 20000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.