missing translation for 'onlineSavingsMsg'
Learn More
Learn More
200.7, 6010 - Instrument Performance Check (IPC) Solution 2: Al, Fe, Hg, Li, Mo, Na, Sb, Sn, Ti @ 100; SiO2 @ 500 μg/mL in 20% HCl, tr. HF, VHG™
Description
200.7, 6010 - Instrument Performance Check (IPC) Solution 2: Al, Fe, Hg, Li, Mo, Na, Sb, Sn, Ti @ 100; SiO2 @ 500 μg/mL in 20% HCl, tr. HF
Specifications
Specifications
| Chemical Name or Material | Aluminium/Antimony/Iron/Lithium/Mercury/Molybdenum/Sodium/Tin/Titanium/Silicon dioxide |
| CAS | 7429-90-5, 7440-36-0, 7439-89-6, 7439-93-2, 7439-97-6, 7439-98-7 |
| Methods | ICP |
| Analyte or Component Names | Aluminium/Antimony/Iron/Lithium/Mercury/Molybdenum/Sodium/Tin/Titanium/Silicon dioxide |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HCl |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 100 μg/mL/Antimony 100 μg/mL/Iron 100 μg/mL/Lithium 100 μg/mL/Mercury 100 μg/mL/Molybdenum 100 μg/mL/Sodium 100 μg/mL/Tin 100 μg/mL/Titanium 100 μg/mL/Silicon dioxide 500 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?