missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat BMP-6 Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Nakamura, K. et al. (1999) Exp. Cell Res. 250:351. The ED50 for this effect is 0.15-0.9 μg/mL.
Specifications
Specifications
| Accession Number | Q04906.2 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in HCl. |
| Gene ID (Entrez) | 654 |
| Molecular Weight (g/mol) | Observed: 16-22 kDa, under reducing conditions; Theroretical: 16 kDa |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived rat BMP-6 protein Ser368-His506 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | BMP6, BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, VGR1, VGR-1 |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction