missing translation for 'onlineSavingsMsg'
Learn More

20(S)-Ginsenoside Rh2, TRC

Product Code. 30240977
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240977 25 mg 25mg
30240747 10 mg 10mg
30240764 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240977 Supplier Toronto Research Chemicals Supplier No. TRCG38758525MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78214-33-2
Molecular Formula C36 H62 O8
Molecular Weight (g/mol) 622.87
Synonym β-D-Glucopyranoside, (3β,12β)-12,20-dihydroxydammar-24-en-3-yl, Dammarane, β-D-glucopyranoside deriv., (3β,12β)-12,20-Dihydroxydammar-24-en-3-yl β-D-glucopyranoside, 20(S)-Ginsenoside Rh2, 20(S)-Rh2, 3-O-β-D-Glucopyranosyl-20(S)-protopanaxadiol, Ginsenoside 20-Rh2, Ginsenoside Rh2
IUPAC Name (2R,3R,4S,5S,6R)-2-[[(3S,5R,8R,9R,10R,12R,13R,14R,17S)-12-hydroxy-17-[(2S)-2-hydroxy-6-methylhept-5-en-2-yl]-4,4,8,10,14-pentamethyl-2,3,5,6,7,9,11,12,13,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl]oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CC(=CCC[C@](C)(O)[C@H]1CC[C@]2(C)[C@@H]1[C@H](O)C[C@@H]3[C@@]4(C)CC[C@H](O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)C(C)(C)[C@@H]4CC[C@@]23C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C36H62O8/c1-20(2)10-9-14-36(8,42)21-11-16-35(7)27(21)22(38)18-25-33(5)15-13-26(32(3,4)24(33)12-17-34(25,35)6)44-31-30(41)29(40)28(39)23(19-37)43-31/h10,21-31,37-42H,9,11-19H2,1-8H3/t21-,22+,23+,24-,25+,26-,27-,28+,29-,30+,31-,33-,34+,35+,36-/m0/s1
Formula Weight 622.4445
Chemical Name or Material Ginsenoside Rh2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.