missing translation for 'onlineSavingsMsg'
Learn More

20(S)-Ginsenoside Rh1, TRC

Product Code. 30390572
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390572 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390572 Supplier Toronto Research Chemicals Supplier No. TRCG41017525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ginsenoside Rh1
CAS 63223-86-9
Molecular Formula C36 H62 O9
Quantity 25 mg
Synonym β-D-Glucopyranoside, (3β,6α,12β)-3,12,20-trihydroxydammar-24-en-6-yl, Dammarane, β-D-glucopyranoside deriv., (3β,6α,12β)-3,12,20-Trihydroxydammar-24-en-6-yl β-D-glucopyranoside, 20(S)-Ginsenoside Rh1, Ginsenoside Rh1, Prosapogenin A2, S-Ginsenoside Rh1, Sanchinoside B2, Sanchinoside Rh1
InChI Formula InChI=1S/C36H62O9/c1-19(2)10-9-13-36(8,43)20-11-15-34(6)26(20)21(38)16-24-33(5)14-12-25(39)32(3,4)30(33)22(17-35(24,34)7)44-31-29(42)28(41)27(40)23(18-37)45-31/h10,20-31,37-43H,9,11-18H2,1-8H3/t20-,21+,22-,23+,24+,25-,26-,27+,28-,29+,30-,31+,33+,34+,35+,36-/m0/s1
SMILES CC(=CCC[C@](C)(O)[C@H]1CC[C@]2(C)[C@@H]1[C@H](O)C[C@@H]3[C@@]4(C)CC[C@H](O)C(C)(C)[C@@H]4[C@H](C[C@@]23C)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)C
IUPAC Name (2R,3R,4S,5S,6R)-2-[[(3S,5R,6S,8R,9R,10R,12R,13R,14R,17S)-3,12-dihydroxy-17-[(2S)-2-hydroxy-6-methylhept-5-en-2-yl]-4,4,8,10,14-pentamethyl-2,3,5,6,7,9,11,12,13,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-6-yl]oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 638.87
Formula Weight 638.4394
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.