missing translation for 'onlineSavingsMsg'
Learn More

20(R)-Ginsenoside Rh2, MedChemExpress

Product Code. p-200100066
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16471628

Brand: Medchem Express HYN1401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 20(R)-Ginsenoside Rh2, a matrix metalloproteinase (MMP) inhibitor, acts as a cell antiproliferator.

It has anticancer effects via blocking cell proliferation and causing G1 phase arrest. 20(R)-Ginsenoside Rh2 induces apoptosis, and has anti-inflammatory and antioxidative activity. 20(R)-Ginsenoside Rh2 inhibits the replication and proliferation of mouse and human gammaherpesvirus 68 (MHV-68) with an IC50 of 2.77 μM for murine MHV-68.

Specifications

Chemical Name or Material 20(R)-Ginsenoside Rh2
Physical Form Solid
CAS 112246-15-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C36H62O8
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 125 mg/mL (200.68 mM; Need ultrasonic)
SMILES O[C@H]([C@H]([C@@H]([C@@H](CO)O1)O)O)[C@@H]1O[C@H]2CC[C@]3(C)[C@@]4([H])C[C@@H](O)[C@@H]5[C@@H]([C@](C)(O)CC/C=C(C)\C)CC[C@](C)5[C@@](C)4CC[C@@]3([H])C2(C)C
Molecular Weight (g/mol) 622.87
Formula Weight 622.87
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.