missing translation for 'onlineSavingsMsg'
Learn More

20(R)-Ginsenoside Rh2, MedChemExpress

Product Code. 16471628
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471628 5 mg 5mg
16439218 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471628 Supplier Medchem Express Supplier No. HYN1401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 20(R)-Ginsenoside Rh2, a matrix metalloproteinase (MMP) inhibitor, acts as a cell antiproliferator.

It has anticancer effects via blocking cell proliferation and causing G1 phase arrest. 20(R)-Ginsenoside Rh2 induces apoptosis, and has anti-inflammatory and antioxidative activity. 20(R)-Ginsenoside Rh2 inhibits the replication and proliferation of mouse and human gammaherpesvirus 68 (MHV-68) with an IC50 of 2.77 μM for murine MHV-68.

Specifications

Chemical Name or Material 20(R)-Ginsenoside Rh2
Physical Form Solid
CAS 112246-15-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C36H62O8
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 125 mg/mL (200.68 mM; Need ultrasonic)
SMILES O[C@H]([C@H]([C@@H]([C@@H](CO)O1)O)O)[C@@H]1O[C@H]2CC[C@]3(C)[C@@]4([H])C[C@@H](O)[C@@H]5[C@@H]([C@](C)(O)CC/C=C(C)\C)CC[C@](C)5[C@@](C)4CC[C@@]3([H])C2(C)C
Molecular Weight (g/mol) 622.87
Formula Weight 622.87
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.