missing translation for 'onlineSavingsMsg'
Learn More

20-Hydroxydexamethasone, TRC

Product Code. 30398941
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398941 1 g 1g
30395320 500 mg -
30399823 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398941 Supplier Toronto Research Chemicals Supplier No. TRCH9180951G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55879-48-6
Molecular Formula C22H31FO5
Molecular Weight (g/mol) 394.48
Synonym Pregna-1,4-dien-3-one, 9-fluoro-11,17,20,21-tetrahydroxy-16-methyl-, (11β,16α)- (9CI), 20-Dihydrodexamethasone, 20-Hydroxydexamethasone
IUPAC Name (8S,9R,10S,11S,13S,14S,16R,17R)-17-(1,2-dihydroxyethyl)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(O)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H31FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-18,24,26-28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,18?,19+,20+,21+,22+/m1/s1
Formula Weight 394.2156
Chemical Name or Material 20-Hydroxydexamethasone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.