missing translation for 'onlineSavingsMsg'
Learn More

20-Hydroxydexamethasone, TRC

Product Code. 30395320
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395320 500 mg -
30398941 1 g 1g
30399823 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395320 Supplier Toronto Research Chemicals Supplier No. TRCH918095500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55879-48-6
Molecular Formula C22H31FO5
Molecular Weight (g/mol) 394.48
Synonym Pregna-1,4-dien-3-one, 9-fluoro-11,17,20,21-tetrahydroxy-16-methyl-, (11β,16α)- (9CI), 20-Dihydrodexamethasone, 20-Hydroxydexamethasone
IUPAC Name (8S,9R,10S,11S,13S,14S,16R,17R)-17-(1,2-dihydroxyethyl)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(O)CO

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C22H31FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-18,24,26-28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,18?,19+,20+,21+,22+/m1/s1
Formula Weight 394.2156
Chemical Name or Material 20-Hydroxydexamethasone
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.