missing translation for 'onlineSavingsMsg'
Learn More

20-Dihydroprogesterone Acetate, TRC

Product Code. 30381551
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381551 100 mg 100mg
30378609 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381551 Supplier Toronto Research Chemicals Supplier No. TRCD449690100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5062-62-4
Molecular Formula C23 H34 O3
Molecular Weight (g/mol) 358.51
Synonym (20R)-3-Oxopregn-4-en-20-yl Acetate, Progesterone Imp. E (EP), Pregn-4-en-3-one, 20-(acetyloxy)-, (20R)- (9CI), Pregn-4-en-3-one, 20β-hydroxy-, acetate (6CI,7CI,8CI), 20-Acetoxy-3-oxopregn-4-ene, 20-Dihydroprogesterone acetate, 20β-Acetoxypregn-4-en-3-one
IUPAC Name [(1R)-1-[(8S,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]ethyl] acetate
SMILES C[C@@H](OC(=O)C)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C23H34O3/c1-14(26-15(2)24)19-7-8-20-18-6-5-16-13-17(25)9-11-22(16,3)21(18)10-12-23(19,20)4/h13-14,18-21H,5-12H2,1-4H3/t14-,18+,19-,20+,21+,22+,23-/m1/s1
Formula Weight 358.2508
Chemical Name or Material (20R)-3-Oxopregn-4-en-20-yl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.