missing translation for 'onlineSavingsMsg'
Learn More

20 cSt Mineral Oil Blank - Trace metals and sulfur reported, VHG™

Product Code. 30003043
Click to view available options
Quantity:
500 mL
0.5 gal
1 gal
Unit Size:
0.5 GAL
1 GAL
500mL
This item is not returnable. View return policy

Product Code. 30003043

Brand: VHG Labs VHGOIL20MIN1/2GAL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

20 cSt Mineral Oil Blank - Trace metals and sulfur reported
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 20 cSt Mineral Oil Blank
Methods ICP/XRF/RDE
Analyte or Component Names 20 cSt Mineral Oil Blank
Shipping Condition Room Temperature
IUPAC Name 20 cSt Mineral Oil Blank
Solvent or Matrix 20 cSt Mineral Oil
Quantity 0.5 gal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.