missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylnaphthalene, Stabilized 98%, ACROS Organics™

Brand:  Acros Organics 140900050

Code : 29

Additional Details : CAS Number : 827-54-3 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10306550

  • £35.40 / 5g


EU Stock 6
Add to basket

Specifications

Chemical Identifiers

2-Vinylnaphthalene
827-54-3
100.0
97%
C10H7CH=CH2
MFCD00004125
98%
96.0
≥96.0%
C12H10
154.21

Specifications

2-Vinylnaphthalene
827-54-3
100.0
Glass bottle
≥96.0%
0.5% max. p-tert.-butylcatechol
C10H7CH=CH2
MFCD00004125
63° to 68°C
98%
96.0
Authentic
Powder
97%
C12H10
154.21
White to slightly beige or yellow
5g
Videos
SDS