missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Vinylanthracene, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 2-Vinylanthracene |
| CAS | 2026-16-6 |
| Molecular Formula | C16 H12 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C16H12/c1-2-12-7-8-15-10-13-5-3-4-6-14(13)11-16(15)9-12/h2-11H,1H2 |
| SMILES | C=Cc1ccc2cc3ccccc3cc2c1 |
| IUPAC Name | 2-ethenylanthracene |
| Molecular Weight (g/mol) | 204.27 |
| Formula Weight | 204.0939 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction