missing translation for 'onlineSavingsMsg'
Learn More

Vinylboronic acid pinacol ester, 97+%, stabilized with 0.05% BHT

Product Code. 10400744 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10400744 5 g Glass bottle 5g
10430554 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10400744 Supplier Thermo Scientific Acros Supplier No. 395530050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75927-49-0 | C8H15BO2 | 154.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75927-49-0
Molecular Formula C8H15BO2
Molecular Weight (g/mol) 154.02
MDL Number MFCD00192492
InChI Key DPGSPRJLAZGUBQ-UHFFFAOYSA-N
Synonym vinylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-vinyl-1,3,2-dioxaborolane, vinylboronic acid pinacol cyclic ester, 2-vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-ethenyl-4,4,5,5-tetramethyl, pinacol vinylboronate, stabilized with bht, 2-vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaoborolane, vbpe, pubchem23743, ethenylboronic acid pinacol ester
PubChem CID 5233012
IUPAC Name 2-ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C=C

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color White to Yellow
Density 0.9080g/mL
Boiling Point 40°C (20.0 mmHg)
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4240 to 1.4260 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.908
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Stabilizer 500ppm (BHT) max.
Formula Weight 154.02
Percent Purity 97+%
Physical Form Powder or Low Melting Solid
Chemical Name or Material Vinylboronic acid pinacol ester
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.