missing translation for 'onlineSavingsMsg'
Learn More

2-Undecanone, 98%

Product Code. 11483228
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483228 100 g 100g
11493228 500 g 500g
11403238 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483228 Supplier Thermo Scientific Alfa Aesar Supplier No. A17659.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-12-9 | C11H22O | 170.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-12-9
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.30
MDL Number MFCD00009583
InChI Key KYWIYKKSMDLRDC-UHFFFAOYSA-N
Synonym 2-undecanone, methyl nonyl ketone, 2-hendecanone, ketone, methyl nonyl, undecanone, nonyl methyl ketone, rue ketone, 2-oxoundecane, methyl n-nonyl ketone, methylnonylketone
PubChem CID 8163
ChEBI CHEBI:17700
IUPAC Name undecan-2-one
SMILES CCCCCCCCCC(C)=O

Specifications

Melting Point 11°C to 13°C
Boiling Point 228°C
Flash Point 89°C (192°F)
Quantity 100 g
UN Number UN3082
Beilstein 1749573
Merck Index 14,6104
Formula Weight 170.3
Percent Purity 98%
Chemical Name or Material 2-Undecanone
compliance-icons
Product Identifier
  • 2-Undecanone
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-Undecanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.