missing translation for 'onlineSavingsMsg'
Learn More

2-Undecanol, TRC

Product Code. 30134150
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134150 10 g 10g
30133483 25 g 25g
30012396 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134150 Supplier Toronto Research Chemicals Supplier No. TRCU78881510G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1653-30-1
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.31
Synonym sec-Undecanol, 2-Hendecanol, 2-Hydroxyundecane, Methyl Nonyl Carbinol, sec-Undecyl Alcohol, alpha-Methyldecanol
IUPAC Name undecan-2-ol
SMILES CCCCCCCCCC(C)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H24O/c1-3-4-5-6-7-8-9-10-11(2)12/h11-12H,3-10H2,1-2H3
Formula Weight 172.1827
Chemical Name or Material 2-Undecanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.