missing translation for 'onlineSavingsMsg'
Learn More

2-Undecanol, 98+%

Product Code. 11467087
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467087 50 g 50g
11457087 10 g 10g
11477087 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467087 Supplier Thermo Scientific Alfa Aesar Supplier No. A14441.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1653-30-1 | C11H24O | 172.31 g/mol

It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1653-30-1
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.31
MDL Number MFCD00021958
InChI Key XMUJIPOFTAHSOK-UHFFFAOYNA-N
Synonym 2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol
PubChem CID 15448
ChEBI CHEBI:77930
IUPAC Name undecan-2-ol
SMILES CCCCCCCCCC(C)O

Specifications

Melting Point 1°C to 3°C
Density 0.828
Boiling Point 228°C to 229°C
Flash Point 107°C (225°F)
Odor Mild
Refractive Index 1.437
Quantity 50 g
Beilstein 1719795
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether.
Formula Weight 172.31
Percent Purity ≥98%
Chemical Name or Material 2-Undecanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.