missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)thiazole, 97%

Product Code. 11444977
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444977 5 g 5g
11434977 1 g 1g
11454977 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444977 Supplier Thermo Scientific Alfa Aesar Supplier No. B21903.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79265-30-8 | C6H11NSSi | 157.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79265-30-8
Molecular Formula C6H11NSSi
Molecular Weight (g/mol) 157.31
MDL Number MFCD00066274
InChI Key VJCHUDDPWPQOLH-UHFFFAOYSA-N
Synonym 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq
PubChem CID 588453
IUPAC Name trimethyl(1,3-thiazol-2-yl)silane
SMILES C[Si](C)(C)C1=NC=CS1

Specifications

Density 0.985
Boiling Point 53°C to 55°C (10 mmHg)
Flash Point 57°C (135°F)
Refractive Index 1.497
Quantity 5 g
UN Number UN1993
Sensitivity Moisture sensitive
Formula Weight 157.31
Percent Purity 97%
Chemical Name or Material 2-(Trimethylsilyl)thiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.