missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)thiazole, 96%

Product Code. 10731721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10731721 5 mL Glass bottle 5mL
10073813 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10731721 Supplier Thermo Scientific Acros Supplier No. 292290050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 79265-30-8 | C6H11NSSi | 157.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79265-30-8
Molecular Formula C6H11NSSi
Molecular Weight (g/mol) 157.31
MDL Number MFCD00066274
InChI Key VJCHUDDPWPQOLH-UHFFFAOYSA-N
Synonym 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq
PubChem CID 588453
IUPAC Name trimethyl(1,3-thiazol-2-yl)silane
SMILES C[Si](C)(C)C1=NC=CS1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 53.0°C to 55.0°C (10.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.5000
Quantity 5 mL
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble
Formula Weight 157.31
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilyl)thiazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.