missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)pyridine, 97%, Thermo Scientific™

Product Code. 11367079
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367079 10 g 10g
11357079 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367079 Supplier Thermo Scientific Alfa Aesar Supplier No. H26885.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13737-04-7
Molecular Formula C8H13NSi
Molecular Weight (g/mol) 151.284
MDL Number MFCD06797181
InChI Key IBLKFJNZUSUTPB-UHFFFAOYSA-N
Synonym 2-trimethylsilyl pyridine, pyridine, 2-trimethylsilyl, 2-pyridyltrimethylsilane, 2-trimethylsilylpyridine, 2-pyridyl trimethylsilane, trimethyl pyridin-2-yl silane, amtsi028, acmc-1cb48, trimethyl 2-pyridinyl silane
PubChem CID 139573
IUPAC Name trimethyl(pyridin-2-yl)silane
SMILES C[Si](C)(C)C1=CC=CC=N1

Specifications

Density 0.911
Boiling Point 47°C to 49°C (5 mmHg)
Flash Point 52°C (126°F)
Refractive Index 1.489
Quantity 10 g
UN Number UN1993
Sensitivity Light sensitive
Formula Weight 151.28
Percent Purity 97%
Chemical Name or Material 2-(Trimethylsilyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.